Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_015348.1 | 5 | 1212 | |||
GenBank mRNA | AK133929.1 | - | 3 | 433 | ||
U20366.1 | - | LIT1565 Hoxa11as |
5 | 1212 | ||
U20367.1 | - | LIT1564 Hoxa11as |
3 | 780 | ||
U20368.1 | - | LIT1566 Hoxa11as |
1 | 452 | ||
U20369.1 | - | LIT1563 Hoxa11as |
3 | 417 |