Gene Transcripts
- Gene Symbol
- Phf21a
- Entrez Gene ID
- 192285
- Gene Name
- PHD finger protein 21A
- Gene Aliases
- 80kDa, Bhc, Bhc80, D030065N23Rik, PFTF1
- Chromosome Location
- Chr. 2: 92183746 - 92364667 on Build GRCm38
- UniGene ID
- Mm.330408
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001109690.1 |
NP_001103160.1
|
3, 4 |
145 |
|
NM_001109691.1 |
NP_001103161.1
|
7, 8 |
553 |
|
NM_138755.2 |
NP_620094.2
|
6, 7 |
559 |
|
GenBank mRNA |
AB105178.1 |
- |
6, 7 |
465 |
|
AK037174.1 |
- |
7, 8 |
554 |
|
AK129424.1 |
- |
5, 6 |
1563 |
|
AK137085.1 |
- |
2, 3 |
106 |
|
BC019181.1 |
- |
6, 7 |
414 |
5008954 |
BY303333.1 |
- |
5, 6 |
329 |
|
CX217243.1 |
- |
6, 7 |
559 |
|
View Full Product Details