Gene Transcripts

Gene Symbol
Aldh4a1
Entrez Gene ID
212647
Gene Name
aldehyde dehydrogenase 4 family, member A1
Gene Aliases
A930035F14Rik, Ahd-1, Ahd1, Aldh4, Aldh5a1, E330022C09, P5cd, P5cdh, P5cdhl, P5cdhs, Ssdh1
Chromosome Location
Chr. 4: 139622871 - 139649691 on Build GRCm38
UniGene ID
Mm.273571
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175438.4 NP_780647.3 14 1762
GenBank mRNA AK044712.1 - 14 1558
AK195093.1 - 3 238
BC024133.1 - 13 1480
BC026589.1 - 9 974
BC027140.1 - 10 1173
BC039280.1 - 3 326
BC039281.1 - 14 1546 4975491
BC056226.1 - 14 1646 5064715
BC060650.1 - 13 3152 30354228
CD354968.1 - 4 394

View Full Product Details