Gene Transcripts

Gene Symbol
Csnk1g1
Entrez Gene ID
214897
Gene Name
casein kinase 1, gamma 1
Gene Aliases
9130020E21Rik
Chromosome Location
Chr. 9: 65908962 - 66045015 on Build GRCm38
UniGene ID
Mm.325816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173185.2 NP_775277.2 7 1072
GenBank mRNA AK090262.1 - 7 1073
AK149310.1 - 8 1090
BC064645.1 - 6 1088 5363779

View Full Product Details