Gene Transcripts

Gene Symbol
Rfpl4b
Entrez Gene ID
215919
Gene Name
ret finger protein-like 4B
Gene Aliases
100040915, Gm3037, Gm62
Chromosome Location
Chr. 10: 38816682 - 38822938 on Build GRCm38
UniGene ID
Mm.294313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177783.1 NP_001171254.1 2 796

View Full Product Details