Gene Transcripts

Gene Symbol
Zfyve16
Entrez Gene ID
218441
Gene Name
zinc finger, FYVE domain containing 16
Gene Aliases
AI035632, B130024H06Rik, B130031L15, mKIAA0305
Chromosome Location
Chr. 13: 92487104 - 92532710 on Build GRCm38
UniGene ID
Mm.259585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173392.4 NP_775568.1 3 2301
GenBank mRNA AK122241.1 - 3 2282
BC042669.1 - 3 2299 4235024

View Full Product Details