Gene Transcripts

Gene Symbol
Cyp4a29
Entrez Gene ID
230639
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 29
Gene Aliases
Cyp4a29-ps, OTTMUSG00000008612
Chromosome Location
Chr. 4: 115242084 - 115254557 on Build GRCm38
UniGene ID
Mm.466610
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100183.1 NP_001093653.1 9, 10 1213

View Full Product Details