Gene Transcripts

Gene Symbol
Gbx1
Entrez Gene ID
231044
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Gbx-1
Chromosome Location
Chr. 5: 24504426 - 24526848 on Build GRCm38
UniGene ID
Mm.246596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015739.2 NP_056554.1 2 1054
GenBank mRNA AF547996.1 - 2 1054
AY319256.1 - 2 1408

View Full Product Details