Gene Transcripts

Gene Symbol
Krt39
Entrez Gene ID
237934
Gene Name
keratin 39
Gene Aliases
4732494G06Rik, Ka35
Chromosome Location
Chr. 11: 99514293 - 99521338 on Build GRCm38
UniGene ID
Mm.484091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213730.2 NP_998895.2 1 290
GenBank mRNA BC119541.1 - 1 290
BC128563.1 - 1 285

View Full Product Details