Gene Transcripts

Gene Symbol
Dleu7
Entrez Gene ID
239133
Gene Name
deleted in lymphocytic leukemia, 7
Gene Aliases
BC038059
Chromosome Location
Chr. 14: 62276229 - 62292979 on Build GRCm38
UniGene ID
Mm.127330
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173419.2 NP_775595.1 1, 2 460
GenBank mRNA BC038059.1 - 1, 2 453
CK618986.1 - 1, 2 460

View Full Product Details