Gene Transcripts

Gene Symbol
Serpinb13
Entrez Gene ID
241196
Gene Name
serine (or cysteine) peptidase inhibitor, clade B (ovalbumin), member 13
Gene Aliases
5430417G24, HUR7, HURPIN, PI13, headpin
Chromosome Location
Chr. 1: 106980984 - 107001195 on Build GRCm38
UniGene ID
Mm.110886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172852.3 NP_766440.2 4 349
GenBank mRNA AK030670.1 - 4 351
BC117990.1 - 4 339 40104505
BC117991.1 - 5 404

View Full Product Details