Gene Transcripts

Gene Symbol
Pglyrp3
Entrez Gene ID
242100
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
Gm420
Chromosome Location
Chr. 3: 92012477 - 92032672 on Build GRCm38
UniGene ID
Mm.297406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207247.4 NP_997130.2 5 757
GenBank mRNA AY518698.1 - 5 728
BC128291.1 - 5 728 40102537

View Full Product Details