Gene Transcripts

Gene Symbol
Tmprss11f
Entrez Gene ID
243083
Gene Name
transmembrane protease, serine 11f
Gene Aliases
4631408C20, 4732406D01Rik
Chromosome Location
Chr. 5: 86521226 - 86632424 on Build GRCm38
UniGene ID
Mm.106867
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178730.3 NP_848845.1 9 987
GenBank mRNA AK028447.1 - 9 989
AK028587.1 - 9 982
AK029032.1 - 9 949
AK037173.1 - 8 831
BC132629.1 - 8 837 40130906

View Full Product Details