Gene Transcripts

Gene Symbol
Dpy19l1
Entrez Gene ID
244745
Gene Name
dpy-19-like 1 (C. elegans)
Gene Aliases
1100001I19Rik, 6030402K12, AI225861, AW558481
Chromosome Location
Chr. 9: 24411776 - 24503341 on Build GRCm38
UniGene ID
Mm.38526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172920.4 NP_766508.3 10 1031
GenBank mRNA AK143845.1 - 11 1036
BC031382.1 - 3 209

View Full Product Details