Gene Transcripts

Gene Symbol
Cypt2
Entrez Gene ID
245566
Gene Name
cysteine-rich perinuclear theca 2
Gene Aliases
Ckt1, Cypt2-ps
Chromosome Location
Chr. X: 105499772 - 105500639 on Build GRCm38
UniGene ID
Mm.250428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173436.2 NP_775612.2 2 347
GenBank mRNA AF463499.1 - 2 330
BC060953.1 - 2 347 6774045

View Full Product Details