Gene Transcripts

Gene Symbol
Obox5
Entrez Gene ID
252829
Gene Name
oocyte specific homeobox 5
Gene Aliases
C87544
Chromosome Location
Chr. 7: 15750370 - 15759274 on Build GRCm38
UniGene ID
Mm.222738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145709.2 NP_663755.2 2 220
GenBank mRNA BC052664.1 - 2 224
CJ052752.1 - 2 222

View Full Product Details