Gene Transcripts

Gene Symbol
Klk8
Entrez Gene ID
259277
Gene Name
kallikrein related-peptidase 8
Gene Aliases
BSP1, Nrpn, Prss19
Chromosome Location
Chr. 7: 43797552 - 43803822 on Build GRCm38
UniGene ID
Mm.458023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008940.2 NP_032966.1 3 837
GenBank mRNA AB074296.1 - 1 99
BC055895.1 - 3 351 4019947
D30785.1 - 3 837

View Full Product Details