Gene Transcripts

Gene Symbol
Gpr82
Entrez Gene ID
319200
Gene Name
G protein-coupled receptor 82
Gene Aliases
Chromosome Location
Chr. X: 13658153 - 13667433 on Build GRCm38
UniGene ID
Mm.383294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175669.3 NP_783600.1 4 993
GenBank mRNA AK033761.1 - 4 995
BC116704.1 - 4 988 40104936

View Full Product Details