Gene Transcripts

Gene Symbol
Dpep2
Entrez Gene ID
319446
Gene Name
dipeptidase 2
Gene Aliases
F630103D06Rik, MBD-2
Chromosome Location
Chr. 8: 105984945 - 106006348 on Build GRCm38
UniGene ID
Mm.248357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176913.4 NP_795887.2 9 1324
GenBank mRNA AF488552.1 - 9 1324

View Full Product Details