Gene Transcripts
- Gene Symbol
- Jph4
- Entrez Gene ID
- 319984
- Gene Name
- junctophilin 4
- Gene Aliases
- 9330157P13Rik, JP-4, JPHL1
- Chromosome Location
- Chr. 14: 55106826 - 55116935 on Build GRCm38
- UniGene ID
- Mm.19047
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177049.5 |
NP_796023.2
|
2, 3 |
1496 |
|
GenBank mRNA |
AB099712.1 |
- |
2, 3 |
839 |
|
AK031092.1 |
- |
1, 2 |
543 |
|
AK033956.1 |
- |
2, 3 |
1129 |
|
AK034118.1 |
- |
2, 3 |
1130 |
|
AK036161.1 |
- |
2, 3 |
1142 |
|
AK048607.1 |
- |
1, 2 |
597 |
|
AK148135.1 |
- |
1, 2 |
660 |
|
AY341261.1 |
- |
2, 3 |
1496 |
|
AY350744.1 |
- |
1, 2 |
483 |
|
BC052049.1 |
- |
1, 2 |
541 |
5689347 |
BC052376.1 |
- |
2, 3 |
593 |
5696231 |
BC052722.1 |
- |
2, 3 |
567 |
6810400 |
View Full Product Details