Gene Transcripts

Gene Symbol
Rnase9
Entrez Gene ID
328401
Gene Name
ribonuclease, RNase A family, 9 (non-active)
Gene Aliases
ESRL, Rak1
Chromosome Location
Chr. 14: 51038459 - 51041905 on Build GRCm38
UniGene ID
Mm.99528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183032.2 NP_898853.2 2 383
GenBank mRNA AK078986.1 - 2 386
AY226989.1 - 2 384
AY303816.1 - 2 383
BC107218.1 - 2 388 40051762

View Full Product Details