Gene Transcripts
- Gene Symbol
- Fam71f1
- Entrez Gene ID
- 330277
- Gene Name
- family with sequence similarity 71, member F1
- Gene Aliases
- GARI-L1, Gm764, NYD-SP18
- Chromosome Location
- Chr. 6: 29313482 - 29336022 on Build GRCm38
- UniGene ID
- Mm.87624
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289663.1 |
NP_001276592.1
|
2, 3 |
502 |
|
NM_001289664.1 |
NP_001276593.1
|
2, 3 |
502 |
|
NM_001289665.1 |
NP_001276594.1
|
2, 3 |
497 |
|
NM_207258.3 |
NP_997141.2
|
2, 3 |
502 |
|
GenBank mRNA |
AB473725.1 |
- |
2, 3 |
443 |
|
AK133908.1 |
- |
2, 3 |
503 |
|
BC147513.1 |
- |
2, 3 |
485 |
9056505 |
BC172074.1 |
- |
2, 3 |
485 |
9054360 |
BC172085.1 |
- |
2, 3 |
480 |
|
BC172086.1 |
- |
2, 3 |
485 |
9054372 |
View Full Product Details