Gene Transcripts

Gene Symbol
Cmtm1
Entrez Gene ID
100504164
Gene Name
CKLF-like MARVEL transmembrane domain containing 1
Gene Aliases
CHLFH1a, CKLFH, CKLFH1, Cklfsf1
Chromosome Location
Chr. 8: 104293542 - 104310145 on Build GRCm38
UniGene ID
Mm.443302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181990.2 NP_871790.2 1 567
GenBank mRNA AY241873.1 - 344
AY241874.1 - 778
BC145587.1 - 1 567 9054153

View Full Product Details