Gene Transcripts

Gene Symbol
March4
Entrez Gene ID
381270
Gene Name
membrane-associated ring finger (C3HC4) 4
Gene Aliases
BC056494, mKIAA1399
Chromosome Location
Chr. 1: 72427112 - 72536564 on Build GRCm38
UniGene ID
Mm.337023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045533.1 NP_001038998.1 4 2322
GenBank mRNA AK122502.1 - 4 2685
BC056494.1 - 2 225
BC098191.1 - 4 2322 6832500

View Full Product Details