Gene Transcripts

Gene Symbol
Vmn2r17
Entrez Gene ID
384221
Gene Name
vomeronasal 2, receptor 17
Gene Aliases
EG384221
Chromosome Location
Chr. 5: 109420013 - 109453387 on Build GRCm38
UniGene ID
Mm.469879
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104628.1 NP_001098098.1 2 377

View Full Product Details