Gene Transcripts

Gene Symbol
Vmn1r59
Entrez Gene ID
404284
Gene Name
vomeronasal 1 receptor 59
Gene Aliases
V1rd10
Chromosome Location
Chr. 7: 5453827 - 5454759 on Build GRCm38
UniGene ID
Mm.377928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207543.1 NP_997426.1 1 628
GenBank mRNA BC114347.2 - 1 622
BC148233.1 - 707

View Full Product Details