Gene Transcripts

Gene Symbol
Mrpl42
Entrez Gene ID
67270
Gene Name
mitochondrial ribosomal protein L42
Gene Aliases
2700009F22Rik, 2900055D03Rik, D10Ertd322e, HSPC204, L31mt, MRP-L31, PTD007, Rpml31
Chromosome Location
Chr. 10: 95480805 - 95501992 on Build GRCm38
UniGene ID
Mm.30008
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026065.3 NP_080341.1 5 350
GenBank mRNA AK002530.1 - 5 335
AK003427.2 - 5 351
AK013693.1 - 5 347
BC024337.1 - 5 331 4984291

View Full Product Details