Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000990.4 | NP_000981.1 | 4 | 558 | |
GenBank mRNA | AK311767.1 | - | 4 | 214 | |
BC005326.1 | - | 1 | 214 | 4052417 | |
BC020169.1 | - | 3 | 190 | ||
BG687024.1 | - | 248 | |||
BQ058236.1 | - | 4 | 236 | ||
U14968.1 | - | 4 | 208 |