Gene Transcripts

Gene Symbol
Ighv14-4
Entrez Gene ID
629826
Gene Name
immunoglobulin heavy variable 14-4
Gene Aliases
Gm16841, VH-D-J
Chromosome Location
Chr. 12: 114176438 - 114176867 on Build GRCm38
UniGene ID
Mm.425779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M94020.1 - 91

View Full Product Details