Gene Transcripts

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
Chromosome Location
Chr. 11: 112086824 - 112095801 on Build GRCh38
UniGene ID
Hs.356270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276504.1 NP_001263433.1 2 219
NM_001276506.1 NP_001263435.1 3 336
NM_003002.3 NP_002993.1 3 336
NR_077060.1 3 336
GenBank mRNA AB006202.1 - 3 263
AK075360.1 - 3 262
BC005263.1 - 3 280 3930691
BC009574.1 - 3 261 4063808
BC012603.1 - 3 265 4108104
BC015188.1 - 3 287 4279214
BC015992.1 - 3 283 4292983
BC022350.1 - 3 275 4275516
BC070307.1 - 3 261 4277540
BC071755.1 - 3 279 4774525
BC071756.1 - 3 261 6450992
BF211358.1 - 2 144
BG492859.1 - 3 263
BQ932668.1 - 3 344
BT007238.1 - 3 252
CR456932.1 - 3 252
KU178325.1 - 3 252

View Full Product Details