Gene Transcripts
- Gene Symbol
- ALK
- Entrez Gene ID
- 238
- Gene Name
- anaplastic lymphoma receptor tyrosine kinase
- Gene Aliases
- CD246, NBLST3
- Chromosome Location
- Chr. 2: 29192774 - 29921611 on Build GRCh38
- UniGene ID
- Hs.654469
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004304.4 |
NP_004295.2
|
7, 8 |
2509 |
|
GenBank mRNA |
AB209477.4 |
- |
7, 8 |
2046 |
|
EU788003.1 |
- |
7, 8 |
2464 |
|
EU788004.1 |
- |
7, 8 |
2464 |
|
EU788005.1 |
- |
7, 8 |
2464 |
|
EU788006.1 |
- |
7, 8 |
2464 |
|
EU788007.1 |
- |
7, 8 |
2464 |
|
EU788008.1 |
- |
7, 8 |
2464 |
|
EU788009.1 |
- |
7, 8 |
2464 |
|
EU788010.1 |
- |
7, 8 |
2464 |
|
FJ838786.1 |
- |
7, 8 |
2464 |
|
FJ838787.1 |
- |
7, 8 |
2464 |
|
GU128152.1 |
- |
7, 8 |
1557 |
|
GU128153.1 |
- |
7, 8 |
1557 |
|
GU128154.1 |
- |
7, 8 |
1557 |
|
GU128155.1 |
- |
7, 8 |
1557 |
|
GU128156.1 |
- |
7, 8 |
1557 |
|
GU128157.1 |
- |
7, 8 |
1557 |
|
U62540.1 |
- |
7, 8 |
2468 |
|
U66559.1 |
- |
7, 8 |
1705 |
|
View Full Product Details