Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
6442
Gene Name
sarcoglycan alpha
Gene Aliases
50DAG, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin
Chromosome Location
Chr. 17: 50165517 - 50175932 on Build GRCh38
UniGene ID
Hs.463412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000023.3 NP_000014.1 6, 7 801
XM_011525120.2 XP_011523422.2 6, 7 1267
XM_011525124.2 XP_011523426.1 5, 6 622
GenBank mRNA AK290622.1 - 6, 7 774
BC025702.1 - 6, 7 773 5222483
KU178331.1 - 6, 7 745
L34355.1 - 6, 7 782
U08895.1 - 6, 7 756

View Full Product Details