Gene Transcripts

Gene Symbol
Crybb3
Entrez Gene ID
64349
Gene Name
crystallin, beta B3
Gene Aliases
Chromosome Location
Chr. 12: 49564996 - 49571006 on Build Rnor_6.0
UniGene ID
Rn.19693
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031690.1 NP_113878.1 4 432
XM_006249572.1 XP_006249634.1 5 470
GenBank mRNA AF287304.1 - 4 404
X05899.1 - 4 432

View Full Product Details