Gene Transcripts

Gene Symbol
Msx3
Entrez Gene ID
114504
Gene Name
msh homeobox 3
Gene Aliases
Chromosome Location
Chr. 1: 212514223 - 212517151 on Build Rnor_6.0
UniGene ID
Rn.92366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053712.1 NP_446164.1 2 403
GenBank mRNA AB197924.1 - 1 60
AF390078.1 - 1 31

View Full Product Details