Gene Transcripts

Gene Symbol
Pcdhga11
Entrez Gene ID
252897
Gene Name
protocadherin gamma subfamily A, 11
Gene Aliases
Chromosome Location
Chr. 18: 30904364 - 30971113 on Build Rnor_6.0
UniGene ID
Rn.8628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037153.1 NP_001032230.1 1 1227
GenBank mRNA AY574010.1 - 1 1227

View Full Product Details