Gene Transcripts

Gene Symbol
Camta2
Entrez Gene ID
287462
Gene Name
calmodulin binding transcription activator 2
Gene Aliases
Chromosome Location
Chr. 10: 57291192 - 57309655 on Build Rnor_6.0
UniGene ID
Rn.13745
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105801.2 NP_001099271.2 5, 6 451
XM_006246665.2 XP_006246727.1 5, 6 808

View Full Product Details