Gene Transcripts

Gene Symbol
Lpo
Entrez Gene ID
287610
Gene Name
lactoperoxidase
Gene Aliases
Chromosome Location
Chr. 10: 75100385 - 75120320 on Build Rnor_6.0
UniGene ID
Rn.60583
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105829.1 NP_001099299.1 7 1066
XM_006247085.2 XP_006247147.1 7 1139

View Full Product Details