Gene Transcripts

Gene Symbol
Rfc4
Entrez Gene ID
288003
Gene Name
replication factor C (activator 1) 4
Gene Aliases
Chromosome Location
Chr. 11: 81358592 - 81373044 on Build Rnor_6.0
UniGene ID
Rn.17046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105869.2 NP_001099339.1 7 667
GenBank mRNA BC168166.1 - 7 667
FQ228232.1 - 7 699

View Full Product Details