Gene Transcripts

Gene Symbol
Arhgap31
Entrez Gene ID
288093
Gene Name
Rho GTPase activating protein 31
Gene Aliases
Cdgap
Chromosome Location
Chr. 11: 64600968 - 64714114 on Build Rnor_6.0
UniGene ID
Rn.225416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105879.1 NP_001099349.1 2 344

View Full Product Details