Gene Transcripts

Gene Symbol
Rec8
Entrez Gene ID
290227
Gene Name
REC8 meiotic recombination protein
Gene Aliases
Rec8L1
Chromosome Location
Chr. 15: 34296922 - 34304387 on Build Rnor_6.0
UniGene ID
Rn.108612
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011916.1 NP_001011916.1 10, 11 1061
GenBank mRNA AY529701.1 - 10, 11 1077
BC079023.1 - 10, 11 1061 7113870

View Full Product Details