Gene Transcripts

Gene Symbol
Ushbp1
Entrez Gene ID
290629
Gene Name
Usher syndrome 1C binding protein 1
Gene Aliases
Chromosome Location
Chr. 16: 19781276 - 19791832 on Build Rnor_6.0
UniGene ID
Rn.107429
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033687.1 NP_001028859.1 8 1224
GenBank mRNA BC101905.1 - 8 1224

View Full Product Details