Gene Transcripts

Gene Symbol
Tmem9b
Entrez Gene ID
293415
Gene Name
TMEM9 domain family, member B
Gene Aliases
RGD1310775
Chromosome Location
Chr. 1: 174360534 - 174380319 on Build Rnor_6.0
UniGene ID
Rn.204216
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106289.1 NP_001099759.1 3, 4 481
XM_006229972.2 XP_006230034.1 3, 4 1772

View Full Product Details