Gene Transcripts

Gene Symbol
Ankrd50
Entrez Gene ID
294988
Gene Name
ankyrin repeat domain 50
Gene Aliases
RGD1311665
Chromosome Location
Chr. 2: 125289071 - 125323528 on Build Rnor_6.0
UniGene ID
Rn.7482
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191606.1 NP_001178535.1 3 1025
XM_008760920.1 XP_008759142.1 4 1973
XM_008760921.1 XP_008759143.1 4 1645
XM_008760922.1 XP_008759144.1 4 1567

View Full Product Details