Gene Transcripts

Gene Symbol
Crygn
Entrez Gene ID
296730
Gene Name
crystallin, gamma N
Gene Aliases
Chromosome Location
Chr. 4: 6900907 - 6909167 on Build Rnor_6.0
UniGene ID
Rn.23610
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235882.2 XP_006235944.1 2 586

View Full Product Details