Gene Transcripts

Gene Symbol
Rint1
Entrez Gene ID
296750
Gene Name
RAD50 interactor 1
Gene Aliases
RGD1560433
Chromosome Location
Chr. 4: 7851602 - 7885445 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235975.2 XP_006236037.1 8, 9 1316

View Full Product Details