Gene Transcripts

Gene Symbol
Ccin
Entrez Gene ID
298392
Gene Name
calicin
Gene Aliases
Chromosome Location
Chr. 5: 59452348 - 59454235 on Build Rnor_6.0
UniGene ID
Rn.109532
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005904.1 NP_001005904.1 1 813
GenBank mRNA BC083832.1 - 1 813

View Full Product Details