Gene Transcripts
- Gene Symbol
- Ube2j2
- Entrez Gene ID
- 298689
- Gene Name
- ubiquitin-conjugating enzyme E2, J2
- Gene Aliases
- Chromosome Location
- Chr. 5: 173372659 - 173387084 on Build Rnor_6.0
- UniGene ID
- Rn.106299
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007655.1 |
NP_001007656.1
|
7, 8 |
988 |
|
XM_006239554.2 |
XP_006239616.1
|
7, 8 |
840 |
|
XM_006239555.2 |
XP_006239617.1
|
8, 9 |
1175 |
|
XM_006239557.2 |
XP_006239619.1
|
6, 7 |
835 |
|
XM_006239558.2 |
XP_006239620.1
|
6, 7 |
677 |
|
XM_008764352.1 |
XP_008762574.1
|
7, 8 |
998 |
|
XM_008764353.1 |
XP_008762575.1
|
7, 8 |
2068 |
|
XM_008764354.1 |
XP_008762576.1
|
8, 9 |
1179 |
|
XM_008764355.1 |
XP_008762577.1
|
8, 9 |
2124 |
|
GenBank mRNA |
BC079115.1 |
- |
7, 8 |
988 |
|
View Full Product Details