Gene Transcripts

Gene Symbol
Wdr72
Entrez Gene ID
300834
Gene Name
WD repeat domain 72
Gene Aliases
RGD1563550
Chromosome Location
Chr. 8: 80965734 - 81125710 on Build Rnor_6.0
UniGene ID
Rn.95019
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135719.1 NP_001129191.1 16 2863

View Full Product Details