Gene Transcripts

Gene Symbol
Glod5
Entrez Gene ID
302554
Gene Name
glyoxalase domain containing 5
Gene Aliases
RGD1564596
Chromosome Location
Chr. X: 15225645 - 15240458 on Build Rnor_6.0
UniGene ID
Rn.19978
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106957.1 NP_001100427.1 3 264
XM_006256706.2 XP_006256768.1 4 317

View Full Product Details