Gene Transcripts

Gene Symbol
Tekt4
Entrez Gene ID
302991
Gene Name
tektin 4
Gene Aliases
RGD1308075, Tek4, Tektin4
Chromosome Location
Chr. 10: 14828642 - 14833818 on Build Rnor_6.0
UniGene ID
Rn.27474
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013965.1 NP_001013987.1 4 948
GenBank mRNA AB194013.1 - 4 833
BC079304.1 - 4 948 7134598

View Full Product Details